1,2-Epoxycyclopentane ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopentene oxide

075520-5G 075520-25G 075520-100G
102541-730EA 52.31 USD
102541-730 102551-966 102551-968
1,2-Epoxycyclopentane ≥95%
1,2-Epoxycyclopentane

Hazard: Flammable/Irritant

Formula: C₅H₈O
MW: 84.12
Boiling Pt: 101-102°C
Density: 0.965
Flash Pt: 10°C
MDL Number: MFCD00005161
CAS Number: 285-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR