1-Cyclohexenylboronic acid pinacol ester ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Cyclohexeneboronic acid pinacol ester, 2-(Cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Cyclohexene-1-boronic acid pinacol ester
075519-1G
075519-5G
102541-724EA
86.15
USD
102541-724
102541-726
1-Cyclohexenylboronic acid pinacol ester ≥95%
1-Cyclohexenylboronic acid pinacol ester
Hazard: Irritant/Store in Freezer
Formula:
C₁₂H₂₁BO₂ |
MDL Number:
MFCD05663845 CAS Number: 141091-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...