Methyl-3-amino-4-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-methylbenzoate

075516-100G 075516-25G
102541-716EA 230.77 USD
102541-716 102833-844
Methyl-3-amino-4-methylbenzoate ≥95%
Methyl 3-amino-4-methylbenzoate

Hazard: Irritant

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00025206
CAS Number: 18595-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR