(S)-(-)-α-Methylbenzylamine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(-)-1-Phenylethylamine, (S)-(-)-α-Phenylethylamine, L(-)-α-Methylbenzylamine

075515-100G 075515-500G
102541-712EA 58.46 USD
102541-712 102664-058
(S)-(-)-α-Methylbenzylamine ≥95%
(S)-(-)-α-Methylbenzylamine

Hazard: Toxic/Corrosive

Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 187 °C
Density: 0.94
Flash Pt: 70 °C
MDL Number: MFCD00064406
CAS Number: 2627-86-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR