Dasatinib ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide

075512-1G 075512-5G
102541-694EA 101.54 USD
102541-694 102541-696
Dasatinib ≥95%
Dasatinib

Hazard: Irritant

Formula: C₂₂H₂₆ClN₇O₂S
MW: 488 g/mol
MDL Number: MFCD11046566
CAS Number: 302962-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR