9-Anthracenemethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 9-Anthrylmethanol, Anthracene-9-methanol

075511-10G 075511-100G
102541-690EA 46.15 USD
102541-690 102541-692
9-Anthracenemethanol ≥95%
9-Anthracenemethanol

Hazard: Irritant

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Boiling Pt: 371
MDL Number: MFCD00001264
CAS Number: 1468-95-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR