1-Bromo-3,5-di-tert-butylbenzene ≥95%
Supplier: Matrix Scientific
075510-25G
075510-100G
102664-054EA
141.54
USD
102664-054
102664-056
1-Bromo-3,5-di-tert-butylbenzene ≥95%
1-Bromo-3,5-di-tert-butylbenzene
Hazard: Irritant/Store in Freezer
Formula:
C₁₄H₂₁Br MW: 269.23 g/mol Melting Pt: 62-66°C |
MDL Number:
MFCD00796945 CAS Number: 22385-77-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...