4-Bromopiperidine hydrobromide ≥95%
Supplier: Matrix Scientific
075508-1G
075508-5G
075508-10G
102541-672EA
55.38
USD
102541-672
102541-674
102541-676
4-Bromopiperidine hydrobromide ≥95%
4-Bromopiperidine hydrobromide
Hazard: Irritant
Formula:
C₅H₁₁Br₂N Melting Pt: 199…202 °C |
MDL Number:
MFCD00191858 CAS Number: 54288-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...