6-(Benzyloxy)-7H-purin-2-amine ≥95%

Supplier: Matrix Scientific

Synonyms: O6-Benzylguanine

075506-10G 075506-100G
102541-662EA 116.92 USD
102541-662 102541-664
6-(Benzyloxy)-7H-purin-2-amine ≥95%
6-(Benzyloxy)-7H-purin-2-amine

Hazard: Irritant

Formula: C12H11N5O
MW: 241.2 g/mol
MDL Number: MFCD00269931
CAS Number: 19916-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR