8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline ≥95%

Supplier: Matrix Scientific

075504-1G 075504-5G 075504-10G
102541-654EA 355.38 USD
102541-654 102541-656 102541-658
8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline ≥95%
8-Bromo-4-hydroxy-2-(trifluoromethyl)quinoline

Hazard: Irritant

Formula: C₁₀H₅BrF₃NO
MW: 292.05 g/mol
MDL Number: MFCD00174291
CAS Number: 59108-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR