5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol ≥95%

Supplier: Matrix Scientific

Synonyms: 5,7-Dichloro-4-hydroxy-2-(trifluoromethyl)quinoline

075503-1G 075503-5G 075503-10G
102541-648EA 355.38 USD
102541-648 102541-650 102541-652
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol ≥95%
5,7-Dichloro-2-(trifluoromethyl)-4-quinolinol

Hazard: Irritant

Formula: C₁₀H₄Cl₂F₃NO
MW: 282.0500 g/mol
MDL Number: MFCD00173851
CAS Number: 59108-13-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR