Methyl-3-bromo-4-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-bromo-4-methylbenzoate

075502-10G 075502-100G
102541-644EA 198.46 USD
102541-644 102541-646
Methyl-3-bromo-4-methylbenzoate ≥95%
Methyl 3-bromo-4-methylbenzoate

Hazard: Irritant

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
MDL Number: MFCD00144769
CAS Number: 104901-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR