4-Amino-6-chloro-2-(methylthio)pyrimidine ≥95%

Supplier: Matrix Scientific

075501-1G 075501-5G 075501-10G
102541-638EA 38.46 USD
102541-638 102541-640 102541-642
4-Amino-6-chloro-2-(methylthio)pyrimidine ≥95%
4-Amino-6-chloro-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₅H₆ClN₃S
MW: 175.6400 g/mol
MDL Number: MFCD00006088
CAS Number: 1005-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR