L(+)-Isoleucinol ≥95%

Supplier: Matrix Scientific

Synonyms: L-Isoleucinol, (S)-(+)-Isoleucinol, (2S,3S)-2-Amino-3-methyl-1-pentanol, (2S,3S)-2-Amino-3-methylpentan-1-ol

075500-5G 075500-25G 075500-100G
102541-634EA 21.54 USD
102541-634 102833-840 102833-842
L(+)-Isoleucinol ≥95%
L(+)-Isoleucinol

Hazard: Irritant

Formula: C₆H₁₅NO
MW: 117.19 g/mol
MDL Number: MFCD00004731
CAS Number: 24629-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR