4-Phenyl-1-(p-tolylsulfonyl)piperidine-4-carbonitrile ≥95%

Supplier: Matrix Scientific

075498-1G 075498-5G 075498-10G
102541-626EA 178.46 USD
102541-626 102541-628 102541-630
4-Phenyl-1-(p-tolylsulfonyl)piperidine-4-carbonitrile ≥95%
4-Phenyl-1-(p-tolylsulfonyl)piperidine-4-carbonitrile

Hazard: Irritant

Formula: C₁₉H₂₀N₂O₂S
CAS Number: 24476-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR