4-Bromo-2,2-diphenylbutanenitrile ≥95%

Supplier: Matrix Scientific

075497-10G 075497-100G
102541-622EA 13.08 USD
102541-622 102541-624
4-Bromo-2,2-diphenylbutanenitrile ≥95%
4-Bromo-2,2-diphenylbutanenitrile

Hazard: Irritant/Refrigerate

Formula: C₁₆H₁₄BrN
MDL Number: MFCD00001845
CAS Number: 39186-58-8
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR