2,3-Dimethylmaleic anhydride ≥95%
Supplier: Matrix Scientific
075493-100G
075493-25G
102541-608EA
110.77
USD
102541-608
102833-838
2,3-Dimethylmaleic anhydride ≥95%
2,3-Dimethylmaleic anhydride
Hazard: Irritant
Formula:
C₆H₆O₃ Melting Pt: 222…223 °C |
MDL Number:
MFCD00005523 CAS Number: 766-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...