Methyl-4-acetylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-acetylbenzoate

075492-100G 075492-25G
102541-602EA 180 USD
102541-602 102833-836
Methyl-4-acetylbenzoate ≥95%
Methyl 4-acetylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₀O₃
Melting Pt: 92…96 °C
MDL Number: MFCD00216474
CAS Number: 3609-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR