Bis(dibenzylideneacetone)palladium ≥95%

Supplier: Matrix Scientific

Synonyms: Bis(dibenzylideneacetone)palladium(0)

075490-1G 075490-5G 075490-25G
102541-588EA 69.23 USD
102541-588 102541-590 102551-964
Bis(dibenzylideneacetone)palladium ≥95%
Bis(dibenzylideneacetone)palladium

Hazard: Irritant/Air Sensitive/Moisture Sensitive/Stored under Argon

Formula: C₃₄H₂₈O₂Pd
Melting Pt: 135 °C (dec.)
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR