1,2,4-Trimethoxybenzene ≥95%
Supplier: Matrix Scientific
075489-10G
075489-100G
102541-584EA
64.62
USD
102541-584
102541-586
1,2,4-Trimethoxybenzene ≥95%
1,2,4-Trimethoxybenzene
Hazard: Irritant
Formula:
C₉H₁₂O₃ MW: 168.19 g/mol |
MDL Number:
MFCD00008360 CAS Number: 135-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...