4'-Hydroxyvalerophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Hydroxyphenyl)-1-pentanone

075488-10G 075488-100G
102541-580EA 64.62 USD
102541-580 102541-582
4'-Hydroxyvalerophenone ≥95%
4'-Hydroxyvalerophenone

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00009719
CAS Number: 2589-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR