4-Chloro-3-nitroanisole ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Chloro-4-methoxy-2-nitrobenzene

075485-10G 075485-100G
102541-568EA 58.46 USD
102541-568 102541-570
4-Chloro-3-nitroanisole ≥95%
4-Chloro-3-nitroanisole

Hazard: Irritant

Formula: C₇H₆ClNO₃
MDL Number: MFCD00007077
CAS Number: 10298-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR