(3-Chlorophenyl)acetic acid ≥95%
Supplier: Matrix Scientific
075480-100G
075480-25G
102541-548EA
75.38
USD
102541-548
102551-962
(3-Chlorophenyl)acetic acid ≥95%
(3-Chlorophenyl)acetic acid
Hazard: Irritant
Formula:
(C₆H₄Cl)CH₂COOH Melting Pt: 77…80 °C |
MDL Number:
MFCD00004332 CAS Number: 1878-65-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...