5-Bromo-1,2,3-trimethoxybenzene ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-3,4,5-trimethoxybenzene
075478-10G
075478-100G
102541-538EA
178.46
USD
102541-538
102541-540
5-Bromo-1,2,3-trimethoxybenzene ≥95%
5-Bromo-1,2,3-trimethoxybenzene
Hazard: Irritant
Formula:
C₉H₁₁BrO₃ MW: 247.09 g/mol |
MDL Number:
MFCD00017169 CAS Number: 2675-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...