3,5-Dimethylphenylhydrazinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(3,5-Dimethylphenyl)hydrazine hydrochloride, 3,5-Dimethylphenylhydrazine hydrochloride

075476-10G 075476-100G
102541-530EA 161.54 USD
102541-530 102541-532
3,5-Dimethylphenylhydrazinium chloride ≥95%
3,5-Dimethylphenylhydrazinium chloride

Hazard: Irritant

Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
MDL Number: MFCD00052269
CAS Number: 60481-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR