2,3-Dimethoxybenzaldehyde ≥95%

Supplier: Matrix Scientific

075475-10G 075475-100G
102541-526EA 15.38 USD
102541-526 102541-528
2,3-Dimethoxybenzaldehyde ≥95%
2,3-Dimethoxybenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₉H₁₀O₃
Boiling Pt: 178 °C (20 mmHg)
Melting Pt: 51…52 °C
MDL Number: MFCD00003309
CAS Number: 86-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR