(R)-(+)-4-Benzyl-2-oxazolidinone ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-4-Benzyl-2-oxazolidinone

075472-100G 075472-25G
102541-514EA 315.38 USD
102541-514 102833-832
(R)-(+)-4-Benzyl-2-oxazolidinone ≥95%
(R)-(+)-4-Benzyl-2-oxazolidinone

Hazard: Irritant/Hygroscopic

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 88-90°C
MDL Number: MFCD00010846
CAS Number: 102029-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR