1-Bromo-2-chlorobenzene ≥95%

Supplier: Matrix Scientific

075465-100G 075465-25G
102541-482EA 138.46 USD
102541-482 102833-830
1-Bromo-2-chlorobenzene ≥95%
1-Bromo-2-chlorobenzene

Hazard: Irritant

Formula: C₆H₄BrCl
MDL Number: MFCD00000532
CAS Number: 694-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR