(S)-1-(Naphthalen-1-yl)ethanol ≥95%

Supplier: Matrix Scientific

075464-1G 075464-5G 075464-10G
102541-474EA 178.46 USD
102541-474 102541-476 102541-478
(S)-1-(Naphthalen-1-yl)ethanol ≥95%
(S)-1-(Naphthalen-1-yl)ethanol

Hazard: Irritant

Formula: C₁₂H₁₂O
MDL Number: MFCD00077831
CAS Number: 15914-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR