(R,R)-(+)-Hydrobenzoin ≥95%

Supplier: Matrix Scientific

Synonyms: (1R,2R)-1,2-Diphenylethane-1,2-diol, (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol

075463-1G 075463-5G 075463-10G
102541-468EA 178.46 USD
102541-468 102541-470 102541-472
(R,R)-(+)-Hydrobenzoin ≥95%
(R,R)-(+)-Hydrobenzoin

Hazard: Irritant

Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
MDL Number: MFCD00064254
CAS Number: 52340-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR