(±)-α-Bromophenylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: α-Bromophenylacetic Acid, DL-α-Bromophenylacetic acid

075460-10G 075460-100G
102541-458EA 90.77 USD
102541-458 102541-460
(±)-α-Bromophenylacetic acid ≥95%
(±)-α-Bromophenylacetic acid

Hazard: Irritant

Formula: C₈H₇BrO₂
MW: 215.0440 g/mol
MDL Number: MFCD00004206
CAS Number: 4870-65-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR