6-Benzylaminopurine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-BAP, 6-Benzyladenine, N6-Benzyladenine, Benzyl(purin-6-yl)amine

075454-10G 075454-100G
102541-432EA 64.62 USD
102541-432 102541-434
6-Benzylaminopurine ≥95%
6-Benzylaminopurine

TCA listed: T
Hazard: Irritant-Harmful/Refrigerate

Formula: C₁₂H₁₁N₅
MW: 225.25 g/mol
MDL Number: MFCD00005572
CAS Number: 1214-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR