1,4-Cyclohexanedione ≥95%

Supplier: Matrix Scientific

075453-10G 075453-100G
102541-428EA 64.62 USD
102541-428 102541-430
1,4-Cyclohexanedione ≥95%
1,4-Cyclohexanedione

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 227
MDL Number: MFCD00001606
CAS Number: 637-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR