N-Methyl-1-(p-tolyl)methanamine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Methyl-4-methylbenzylamine

075450-1G 075450-5G 075450-10G
102541-412EA 18.46 USD
102541-412 102541-414 102541-416
N-Methyl-1-(p-tolyl)methanamine ≥95%
N-Methyl-1-(p-tolyl)methanamine

Hazard: Irritant

Formula: C₉H₁₃N
MW: 135.2100 g/mol
MDL Number: MFCD04115412
CAS Number: 699-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR