Benzoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 1,3-Benzoxazole

075448-10G 075448-100G
102541-406EA 84.62 USD
102541-406 102541-408
Benzoxazole ≥95%
Benzoxazole

Hazard: Irritant

Formula: C₇H₅NO
MW: 119.12 g/mol
MDL Number: MFCD00005765
CAS Number: 273-53-0
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR