1-Boc-3-carbamoylpiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-Carbamoyl-1-piperidinecarboxylate, 1-Boc-3-piperidinecarboxamide, 3-Carbamoyl-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-3-Carbamoylpiperidine, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxamide

075447-1G 075447-5G 075447-10G
102541-400EA 15.38 USD
102541-400 102541-402 102541-404
1-Boc-3-carbamoylpiperidine ≥95%
1-Boc-3-carbamoylpiperidine

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MW: 228.2881 g/mol
MDL Number: MFCD06658365
CAS Number: 91419-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR