Methyl nicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Picolinic acid methyl ester, 3-(Methoxycarbonyl)pyridine, Pyridine-3-carboxylic acid methyl ester, Nicotinic acid methyl ester, 3-Pyridinecarboxylic acid methyl ester, Methyl 3-pyridinecarboxylate
075445-10G
075445-100G
102541-390EA
36.92
USD
102541-390
102541-392
Methyl nicotinate ≥95%
Methyl nicotinate
Hazard: Irritant
Formula:
C₇H₇NO₂ MW: 137.14 g/mol Boiling Pt: 209 Melting Pt: 39 Flash Pt: 95 |
MDL Number:
MFCD00006388 CAS Number: 93-60-7 Merck Index: 12,06174 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...