(2-Nitrophenyl)hydrazine hydrochloride ≥95%

Supplier: Matrix Scientific

075444-10G 075444-100G
102541-386EA 107.69 USD
102541-386 102541-388
(2-Nitrophenyl)hydrazine hydrochloride ≥95%
(2-Nitrophenyl)hydrazine hydrochloride

Hazard: Irritant

Formula: C₆H₈ClN₃O₂
MDL Number: MFCD00060205
CAS Number: 6293-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR