(S)-2-Acetamido-3-methylbutanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: N-Ac-Val-OH, Ac-Val-OH, N-Acetyl-L-Valine

075443-10G 075443-100G
102541-382EA 29.23 USD
102541-382 102541-384
(S)-2-Acetamido-3-methylbutanoic acid ≥95%
(S)-2-Acetamido-3-methylbutanoic acid

Hazard: Irritant

Formula: C₇H₁₃NO₃
MW: 159.2 g/mol
MDL Number: MFCD00066066
CAS Number: 96-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR