Ethyl-N-Cbz-morpholine-2-carboxylate ≥95%

Supplier: Matrix Scientific

075442-250MG 075442-1G 075442-5G
102541-376EA 456.92 USD
102541-376 102541-378 102541-380
Ethyl-N-Cbz-morpholine-2-carboxylate ≥95%
Ethyl-N-Cbz-morpholine-2-carboxylate

Hazard: Irritant

Formula: C₁₅H₁₉NO₅
CAS Number: 1226776-83-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR