3-Hydroxybenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: m-Hydroxybenzonitrile, m-Cyanophenol, 3-Cyanophenol

075439-100G 075439-25G
102541-362EA 196.92 USD
102541-362 102664-050
3-Hydroxybenzonitrile ≥95%
3-Hydroxybenzonitrile

Hazard: Irritant

Formula: C₇H₅NO
MW: 119.12 g/mol
Melting Pt: 78…81 °C
MDL Number: MFCD00002252
CAS Number: 873-62-1
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR