4-Amino-m-cresol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-hydroxytoluene, 4-Amino-3-methylphenol, 1-Amino-4-hydroxy-2-methylbenzene, 4-Hydroxy-2-methylaniline

075435-10G 075435-100G
102541-342EA 47.69 USD
102541-342 102541-344
4-Amino-m-cresol ≥95%
4-Amino-m-cresol

Hazard: Irritant

Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00007871
CAS Number: 2835-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR