4,4',4''-Triaminotriphenylamine ≥95%

Supplier: Matrix Scientific

Synonyms: Tris(4-aminophenyl)amine

075434-1G 075434-5G
102541-336EA 76.92 USD
102541-336 102541-338
4,4',4''-Triaminotriphenylamine ≥95%
4,4',4''-Triaminotriphenylamine

Hazard: Irritant

Formula: C₁₈H₁₈N₄
MW: 290.38 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00778301
CAS Number: 5981-09-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR