m-Tolylhydrazine hydrochloride ≥95%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-(3-Methylphenyl)hydrazine hydrochloride, 3-Tolylhydrazine hydrochloride, m-Tolylhydrazine hydrochloride, m-Tolylhydraziniumchloride, 3-Methylphenylhydrazine hydrochloride, 1-(3-Tolyl)hydrazine hydrochloride, 3-Methylphenylhydrazinium chloride
075432-10G
075432-100G
102541-328EA
364.62
USD
102541-328
102541-330
m-Tolylhydrazine hydrochloride ≥95%
m-Tolylhydrazine hydrochloride
Purity: 95+%
CAS: 637-04-7
MDL Number: MFCD00012932
Molecular Formula: C7H11ClN2
Molecular Weight: 158,63
Hazard: Irritant
Formula:
C₇H₁₀N₂·HCl |
MDL Number:
MFCD00012932 CAS Number: 637-04-7 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...