N-Cbz-L-serine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Carbobenzyloxy-L-serine, Z-Ser-OH, N-Cbz-L-serine

075430-10G 075430-100G
102541-318EA 58.46 USD
102541-318 102541-320
N-Cbz-L-serine ≥95%
N-Cbz-L-serine

Hazard: Irritant

Formula: C₁₁H₁₃NO₅
MW: 239.2 g/mol
MDL Number: MFCD00002662
CAS Number: 1145-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR