(±)-β-Aminophenethyl alcohol ≥95%

Supplier: Matrix Scientific

Synonyms: DL-2-Phenylglycinol, 2-Amino-2-phenylethanol, DL-Phenylglycinol

075427-5G 075427-10G 075427-100G
102541-300EA 56.92 USD
102541-300 102541-302 102541-304
(±)-β-Aminophenethyl alcohol ≥95%
(±)-β-Aminophenethyl alcohol

Hazard: Irritant

Formula: C₈H₁₁NO
MDL Number: MFCD00130145
CAS Number: 7568-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR