Andrographolide ≥95%

Supplier: Matrix Scientific

Synonyms: (S,E)-4-Hydroxy-3-(2-((1R,4aS,5R,6R,8aS)-6-hydroxy-5-(hydroxymethyl)-5,8a-dimethyl-2-methylenedecahydronaphthalen-1-yl)ethylidene)dihydrofuran-2(3H)-one

075423-1G 075423-5G
102541-288EA 15.38 USD
102541-288 102541-290
Andrographolide ≥95%
Andrographolide

Hazard: Irritant

Formula: C₂₀H₃₀O₅
MW: 350.45 g/mol
MDL Number: MFCD07778082
CAS Number: 5508-58-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR