4-Ethynyl-1,1'-biphenyl ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Ethynylbiphenyl, 4-Biphenylacetylene

075417-1G 075417-5G
102541-260EA 61.54 USD
102541-260 102541-262
4-Ethynyl-1,1'-biphenyl ≥95%
4-Ethynyl-1,1'-biphenyl

Hazard: Irritant

Formula: C₁₄H₁₀
MDL Number: MFCD00102191
CAS Number: 29079-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR