N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide ≥95%

Supplier: Matrix Scientific

075415-250MG 075415-1G 075415-5G
102541-248EA 456.92 USD
102541-248 102541-250 102541-252
N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide ≥95%
N-(5-Bromo-2-methylphenyl)-3-cyanobenzamide

Hazard: Irritant

Formula: C₁₅H₁₁BrN₂O
MDL Number: MFCD12719083
CAS Number: 1157457-17-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR