Ethyl-4-bromobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromobenzoic acid ethyl ester, Ethyl 4-bromobenzoate

075412-10G 075412-100G
102541-234EA 47.69 USD
102541-234 102541-236
Ethyl-4-bromobenzoate ≥95%
Ethyl-4-bromobenzoate

Hazard: Irritant

Formula: C₉H₉BrO₂
MDL Number: MFCD00016329
CAS Number: 5798-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR